Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH11X, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106769 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-769 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-769 Supplier Abnova Corporation Supplier No. H00027328A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PCDH11X.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its Y homolog, despite divergence leading to coding region changes, is the most closely related cadherin family member. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with variable cytoplasmic domains. [provided by RefSeq

Sequence: AQASALCYSPPLAQAAAISHSSPLPQVIALHRSQAQSSVSLQQGWVQGADGLCSVDQGVQGSATSQFYTMSERLHPSDDSIKVIPLTTFTPRQQARPSRGDSPIMEEHP

Specifications

Antigen PCDH11X
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PCDH11X.
Formulation 50% glycerol
Gene PCDH11X
Gene Accession No. NM_032969
Gene Alias PCDH-X/PCDH11/PCDHX/PCDHY
Gene Symbols PCDH11X
Host Species Mouse
Immunogen PCDH11X (NP_061850, 1228 a.a. ∽ 1336 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27328
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.