Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH8, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105061 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-061 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-061 Supplier Abnova Corporation Supplier No. H00005100A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PCDH8.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The gene encodes an integral membrane protein that is thought to function in cell adhesion in a CNS-specific manner. Unlike classical cadherins, which are generally encoded by 15-17 exons, this gene includes only 3 exons. Notable is the large first exon encoding the extracellular region, including 6 cadherin domains and a transmembrane region. Alternative splicing yields isoforms with unique cytoplasmic tails. [provided by RefSeq

Sequence: VRYSTFEEDAPGTVIGTLAEDLHMKVSGDTSFRLMKQFNSSLLRVREGDGQLTVGDAGLDRERLCGQAPQCVLAFDVVSFSQEQFRLVH

Specifications

Antigen PCDH8
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PCDH8.
Formulation 50% glycerol
Gene PCDH8
Gene Accession No. NM_002590
Gene Alias ARCADLIN/PAPC
Gene Symbols PCDH8
Host Species Mouse
Immunogen PCDH8 (NP_002581, 32 a.a. ∽ 120 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5100
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.