Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHGB4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105903 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-903 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-903 Supplier Abnova Corporation Supplier No. H00008641A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PCDHGB4.

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. This particular family member is expressed in fibroblasts and is thought to play a role in wound healing in response to injury. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq

Sequence: STAQIHVLVTDANDNAPVFSQDVYRVSLSENVYPGTTVLQVTATDQDEGVNAEITFSFSEASQITQFDLNSNTGEITVLNTLDFEEVKEYSIVLEARDGG

Specifications

Antigen PCDHGB4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PCDHGB4.
Formulation 50% glycerol
Gene PCDHGB4
Gene Accession No. NM_003736
Gene Alias CDH20/FIB2/MGC138293/MGC142167/PCDH-GAMMA-B4
Gene Symbols PCDHGB4
Host Species Mouse
Immunogen PCDHGB4 (NP_003727, 224 a.a. ∽ 323 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8641
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.