Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCGF3 (B01P), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89106291 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-291 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-291 Supplier Abnova Corporation Supplier No. H00010336B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PCGF3 protein.

The protein encoded by this gene contains a C3HC4 type RING finger, which is a motif known to be involved in protein-protein interactions. The specific function of this protein has not yet been determined. [provided by RefSeq

Sequence: MLTRKIKLWDINAHITCRLCSGYLIDATTVTECLHTFCRSCLVKYLEENNTCPTCRIVIHQSHPLQYIGHDRTMQDIVYKLVPGLQEAEMRKQREFYHKLGMEVPGDIKGETCSAKQHLDSHRNGETKADDSSNKEAAEEKPEEDNDYHRSDEQVSICLECNSSKLRGLKRKWIRCSAQATVLHLKKFIAKKLNLSSFNELDILCNEEILGKDHTLKFVVVTRWRFKKAPLLLHYRPKMDLL

Specifications

Antigen PCGF3
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PCGF3 protein.
Formulation PBS with no preservative; pH 7.4
Gene PCGF3
Gene Accession No. NM_006315.4
Gene Alias DKFZp686D20235/DONG1/FLJ36550/FLJ43813/MGC129615/MGC40413/RNF3/RNF3A
Gene Symbols PCGF3
Host Species Mouse
Immunogen PCGF3 (NP_006306.2,1 a.a. ∽ 242 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10336
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.