Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-PD-1, Clone: 16A2.1,

Catalog No. MABF261MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF261MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF261MI Supplier MilliporeSigma™ Supplier No. MABF261
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PD-1 clone: 16A2.1 in Human samples, and it is validated for Flow Cytometry, Western Blotting

Programmed cell death protein 1 (PD-1) (UniProt Q15116), also known as CD279, hPD-1, hPD-I, hSLE1, Protein PD-1, Systemic lupus erythematosus susceptibility 2) is a type I single transmembrane protein encoded by the PDCD1 gene in human. Maintenance of peripheral tolerance by preventing the development of autoimmunity due to T lymphocytes specific for self-antigens is essential for homeostasis of the immune system. PD-1 receptor CD279 and its ligands PD-L1 (B7-H1, CD274) and PD-L2 (B7-DC; CD273) play a vital role in the induction and maintenance of peripheral tolerance. Upregulated PD-1 expression is seen on conventional CD4+ CD25- T cells (Tconv) when exposed to mesenchymal stem cells, resulting in increased Tconv apoptosis due to PD-1 stimulation by the MSC PD-L2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 16A2.1
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q15116
Gene Symbols PDCD1; PD1
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the extracellular domain of human PD-1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_005009
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.