Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-PD-L2, Clone: 24F.10C12,

Catalog No. MABC969 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC969 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC969 Supplier MilliporeSigma™ Supplier No. MABC969
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PD-L2 clone: 24F.10C12 in Human samples, and it is validated for Flow Cytometry, Immunohistochemistry, Neutralization, Western Blotting

Programmed cell death 1 ligand 2 (UniProt Q9BQ51; also known as B7-DC, B7 dendritic cell molecule, B7-H2, Butyrophilin B7-DC, CD273, PD-1 ligand 2, PD-L2, PDCD1 ligand 2, Programmed death ligand 2) is encoded by the PDCD1LG2 (also known as B7DC, CD273, PDCD1L2, PDL2) gene (Gene ID 80380) in human. PD-1 and PD-1 ligands 1&2 (PD-L1 and PD-L2) are B7:CD28 family members that regulate T cell activation and peripheral tolerance. When engaged together with the TCR, the interaction of PD-1 with its ligands delivers an inhibitory signal to T cell proliferation and cytokine production. While PD-L1 is broadly expressed in hematopoietic and nonhematopoietic cells, PD-L2 expression is highly restricted to antigen presenting cells (APCs), including dendritic cells (DCs) and macrophages. The PD-1 pathway plays a key role in the progressive loss of effector T cell responses during chronic HIV infection. Under some conditions, blockade of this pathway is able to restore many T cell functions. PD-L2 is initially produced with signal peptide (a.a. 1-19) sequence, the removal of which yields the mature protien with a large extracellular (a.a. 20-220) region that contains an Ig-like V-type domain (a.a. 21-118) and an Ig-like C2-type domain (a.a. 122-203), followed by a transmembrane domain (a.a. 221-241) and a cytoplasmic tail (a.a. 242-273).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L2
Applications Flow Cytometry, Immunohistochemistry, Neutralization, Western Blot
Classification Monoclonal
Clone 24F.10C12
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing PBS without preservatives.
Gene Accession No. Q9BQ51
Gene Symbols PDCD1LG2; B7DC; CD273; PDCD1L2; PDL2
Host Species Mouse
Immunogen Recombinant human PD-L2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_079515.2
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.