Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD4, Mouse, Clone: k4C1, Abnova™

Catalog No. 89109400 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-400 Supplier Abnova Corporation Supplier No. MAB1123
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant PDCD4.

This gene encodes a protein localized to the nucleus in proliferating cells. Expression of this gene is modulated by cytokines in natural killer and T cells. The gene product is thought to play a role in apoptosis but the specific role has not yet been determined. Two transcripts encoding different isoforms have been identified. [provided by RefSeq

Specifications

Antigen PDCD4
Applications ELISA, Western Blot
Classification Monoclonal
Clone k4C1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant PDCD4.
Dilution Western Blot (1:1000-1:3000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PDCD4
Gene Accession No. NM_014456
Gene Alias H731/MGC33046/MGC33047
Gene Symbols PDCD4
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human PDCD4.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27250
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.