Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDIA3, Mouse, Clone: MaP.ERP57, Abnova™

Catalog No. 89112972 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-972 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-972 Supplier Abnova Corporation Supplier No. MAB6645
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant PDIA3.

This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates. [provided by RefSeq

Specifications

Antigen PDIA3
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MaP.ERP57
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PDIA3.
Dilution Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene PDIA3
Gene Alias ER60/ERp57/ERp60/ERp61/GRP57/GRP58/HsT17083/P58/PI-PLC
Gene Symbols PDIA3
Host Species Mouse
Immunogen Recombinant protein corresponding to human PDIA3.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2923
Test Specificity Detects ∼57KDa.
Target Species Bovine, Canine, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.