Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDPN, Mouse, Clone: 5E2, Abnova™

Catalog No. 89110820 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-820 Supplier Abnova Corporation Supplier No. MAB2099
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant PDPN.

This gene encodes a type-I integral membrane glycoprotein with diverse distribution in human tissues. The physiological function of this protein may be related to its mucin-type character. The homologous protein in other species has been described as a differentiation antigen and influenza-virus receptor. The specific function of this protein has not been determined but it has been proposed as a marker of lung injury. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Specifications

Antigen PDPN
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5E2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant PDPN.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PDPN
Gene Alias AGGRUS/GP36/GP40/Gp38/HT1A-1/OTS8/PA2.26/T1A/T1A-2
Gene Symbols PDPN
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-206 of human PDPN.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10630
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.