Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM1, Mouse, Clone: C31.3, Abnova™

Catalog No. 89109428 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-428 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-428 Supplier Abnova Corporation Supplier No. MAB11259
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against PECAM1.

Specifications

Antigen PECAM1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone C31.3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against PECAM1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) Western Blot (1-3 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene PECAM1
Gene Alias CD31/PECAM-1
Gene Symbols PECAM1
Host Species Mouse
Immunogen Human spleen membranes isolated from a patient with hairy cell leukemia.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5175
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.