Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM1, Mouse, Clone: LCI-4, Abnova™

Catalog No. 89112169 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-169 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-169 Supplier Abnova Corporation Supplier No. MAB5210

Mouse monoclonal antibody raised against partial recombinant PECAM1.

Specifications

Antigen PECAM1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone LCI-4
Conjugate Unconjugated
Description platelet/endothelial cell adhesion molecule
Formulation In tissue culture supernatant (0.09% sodium azide)
Gene PECAM1
Gene Alias CD31, PECAM-1
Gene Symbols PECAM1
Host Species Mouse
Immunogen Recombinant human IgG Fc fusion protein corresponding to pig PECAM1.
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396941
Test Specificity This antibody recognizes porcine CD31, a 140kD cell surface protein also called PECAM-1. CD31 is expressed by various cell types, but particularly by endothelial cells. Clone LCI-4 has been shown to stimulate tyrosine phosphorylation of CD31 on endothelial cells. Cross reacts with human CD31.
Target Species Human, Pig
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.