Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM1, Mouse, PerCP, Clone: MEM-05, Abnova™

Catalog No. 89108445 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-445 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-445 Supplier Abnova Corporation Supplier No. MAB10240
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against of PECAM1.

Specifications

Antigen PECAM1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone MEM-05
Conjugate PerCP
Description Mouse monoclonal antibody raised against of PECAM1.
Dilution Flow Cytometry (10 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene PECAM1
Gene Alias CD31/PECAM-1
Gene Symbols PECAM1
Host Species Mouse
Immunogen Leukocytes of patient suffering from LGL-type leukaemia.
Purification Method Size-exclusion chromatography purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5175
Test Specificity PECAM1, a type I transmembrane glycoprotein expressed on monocytes, platelets, granulocytes, endothelial cells and stem cells of the myeloid lineage.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.