Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-PELP1, Clone: 2F11.1,

Catalog No. MABC303MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC303MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC303MI Supplier MilliporeSigma™ Supplier No. MABC303
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PELP1 clone: 2F11.1 in Human samples, and it is validated for Immunocytochemistry, Western Blotting

Proline-, glutamic acid- and leucine-rich protein 1 (UniProt Q8IZL8; also known as Modulator of non-genomic activity of estrogen receptor, p160, Proline and glutamic acid rich nuclear protein,Transcription factor HMX3) is encoded by the PELP1 (Also known as HMX3, MNAR, P160) gene in human (Gene ID 27043). PELP1 functions as scaffolding protein and mediates protein–protein interactions in cellular processes involving both genomic and extranuclear signaling pathways. Multiple structural domains allow PELP1 to interact with various steroid receptors (SRs) to modulate their activity as either corepressor or coactivator. Dysregulation of PELP1 expression has been observed in breast, ovarian, endometrial, brain, and prostate cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PELP1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2F11.1
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q8IZL8
Gene Symbols PELP1; HMX3; MNAR
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human PELP1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001265170
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.