Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin, Mouse, Clone: Pen 9, Abnova™

Catalog No. 89135913 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-913 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-913 Supplier Abnova Corporation Supplier No. MAB5192
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Penicillin.

Specifications

Antigen Penicillin
Applications ELISA
Classification Monoclonal
Clone Pen 9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Penicillin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Host Species Mouse
Immunogen The benzylpenicilloyl group of penicillium conjugated with transferrin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Pen 9 reacts mainly with the thiazolidin ring of penicillin, but not with the lactam ring. The nature of the side chain in the penicilloyl group does not effect antibody binding as was shown by testing Pen 9 against benzylpenicillin, ampicillin, amoxillin and 6-aminopenicillanic acid. The presence of carrier protein is not essential for the presentation of the antigen associated with Pen 9. Splenocytes were fused with X63-Ag8.653 cells.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.