Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX12, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105088 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-088 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-088 Supplier Abnova Corporation Supplier No. H00005193A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant PEX12.

This gene belongs to the peroxin-12 family. Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene are a cause of Zellweger syndrome (ZWS). [provided by RefSeq

Sequence: MAEHGAHFTAASVADDQPSIFEVVAQDSLMTAVRPALQHVVKVLAESNPTHYGFLWRWFDEIFTLLDLLLQQHYLSRTSASFSENFYGLKRIVMGDTHKSQRLASAGLPKQQLWKSIMFLVLLPYLKVKLEKLVSSLREEDEYSIHPPSSRWKRFYRAFLAAYPFVNMAWEGWFLVQQLRYILGKAQHHSPLLRLAGVQLGRLTVQDIQALEHKPAKASMMQQPARSVSEKINSALKKAVGGVALSLSTGLSVGVFFLQFLDWWYSSENQETIKSLTALPTPPPPVHLDYNSDSPLLPKMKTVCPLCRKTRVNDTVLATSGYVFCYRCVFHYVRSHQACPITGYPTEVQHLIKLYSPEN

Specifications

Antigen PEX12
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant PEX12.
Formulation 50% glycerol
Gene PEX12
Gene Accession No. BC031085
Gene Alias PAF-3
Gene Symbols PEX12
Host Species Mouse
Immunogen PEX12 (AAH31085, 1 a.a. ∽ 359 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5193
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.