Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-Pex3, Clone: 14F3.2,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABC550
Description
Specifically detects Pex3 clone: 14F3.2 in Human samples, and it is validated for Immunohistochemistry, Western Blotting
Pex3 (also known as peroxin-3, peroxisomal assembly protein PEX3, peroxisomal biogenesis factor 3, transformation-related protein 18) is a multitransmembrane peroxisome protein that is encoded by the PEX3 gene in human (also known as PBD10A and TRG18). Pex19 is involved in maintaining peroxisome biosynthesis and integrity by acting as a docking factor for pex19 and, together with other peroxins (PEXs), is essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs), including Zellweger syndrome (ZWS), neonatal adrenoleukodystrophy (NALD), infantile Refsum disease (IRD), and classical rhizomelic chondrodysplasia punctata (RCDP), are lethal diseases characterized by multiple defects in peroxisome function, resulting in failure of importing one or more classes of peroxisomal matrix proteins into the organelle.
Specifications
Pex3 | |
Monoclonal | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
PEX3 | |
GST-tagged recombinant protein corresponding to human Pex3. | |
100 μg | |
Apoptosis, Cancer | |
NP_003621 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG2b κ |
Immunohistochemistry, Western Blot | |
14F3.2 | |
P56589 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction