Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGLYRP3, Mouse, Clone: 187C434, Abnova™

Catalog No. 89107839 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-839 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-839 Supplier Abnova Corporation Supplier No. MAB0068
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of PGLYRP3.

Peptidoglycan recognition proteins, such as PGRPI-alpha, are part of the innate immune system and recognize peptidoglycan, a ubiquitous component of bacterial cell walls.[supplied by OMIM

Specifications

Antigen PGLYRP3
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 187C434
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of PGLYRP3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In ascites (0.05% sodium azide)
Gene PGLYRP3
Gene Alias MGC149197/PGRP-Ialpha/PGRPIA
Gene Symbols PGLYRP3
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 178-193 of human PGLYRP3.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114771
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.