Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGLYRP4, Mouse, Clone: 186C426, Abnova™

Catalog No. 89107841 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-841 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-841 Supplier Abnova Corporation Supplier No. MAB0070
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of PGLYRP4.

Peptidoglycan recognition proteins, such as PGRPI-beta, are part of the innate immune system and recognize peptidoglycan, a ubiquitous component of bacterial cell walls.[supplied by OMIM

Specifications

Antigen PGLYRP4
Applications Western Blot
Classification Monoclonal
Clone 186C426
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of PGLYRP4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene PGLYRP4
Gene Alias PGLYRPIbeta/PGRP-Ibeta/PGRPIB/SBBI67
Gene Symbols PGLYRP4
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 95-110 of human PGLYRP4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57115
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.