Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGR, Mouse, Clone: PR500, Abnova™

Catalog No. 89109490 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-490 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-490 Supplier Abnova Corporation Supplier No. MAB11327
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant PGR.

This gene encodes a member of the steroid receptor superfamily. The encoded protein mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. This gene uses two distinct promotors and translation start sites in the first exon to produce two isoforms, A and B. The two isoforms are identical except for the additional 165 amino acids found in the N-terminus of isoform A only, and mediate their own response genes and physiologic effects with little overlap. The location of transcription initiation for isoform B has not been clearly determined. [provided by RefSeq

Specifications

Antigen PGR
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone PR500
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PGR.
Dilution Western Blot (0.5-1.0ug/ml for 2 hours at Room Temp) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.05% BSA and 0.05% sodium azide)
Gene PGR
Gene Alias NR3C3/PR
Gene Symbols PGR
Host Species Mouse
Immunogen Recombinant human Progesterone Receptor protein
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5241
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.