Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenytoin, Mouse, Clone: 10-8.3.21.1, Abnova™

Catalog No. 89135119 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-119 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-119 Supplier Abnova Corporation Supplier No. MAB2243
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Phenytoin.

Specifications

Antigen Phenytoin
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 10-8.3.21.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Phenytoin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Phenytoin conjugated with KLH.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity 23.4% Cross-Reactivity With p-hydroxy-phenobarbital.
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.