Learn More
PHIP, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PHIP.
Supplier: Abnova Corporation H00055023A01
Description
PHIP binds the pleckstrin homology (PH) domain of insulin receptor substrate-1 (IRS1; MIM 147545), modulates insulin signaling, and plays a role in pancreatic beta cell growth and survival (Farhang-Fallah et al., 2000 [PubMed 11018022]; Podcheko et al., 2007 [PubMed 17636024]).[supplied by OMIM
Sequence: TLSKSSAVIEQGDCKNNALVPGTIQVNGHGGQPSKLVKRGPGRKPKVEVNTNSGEIIHKKRGRKPKKLQYAKPEDLEQNNVHPIRDEVLPSSTCNFLSETNNVKEDLSpecifications
PHIP | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant PHIP. | |
PHIP | |
FLJ20705/FLJ45918/MGC90216/WDR11/ndrp | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_017934 | |
PHIP | |
PHIP (NP_060404, 1599 a.a. ∽ 1705 a.a) partial recombinant protein with GST tag. | |
RUO | |
55023 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only