Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNNI3, Mouse, Clone: 1G11, Abnova™

Catalog No. 89135341 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-341 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-341 Supplier Abnova Corporation Supplier No. MAB3062
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against phospho-TNNI3.

Specifications

Antigen TNNI3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1G11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against phospho-TNNI3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Phosphorylated TNNI3.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This anribody reacts with phosphorylated form of TNNI3.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.