Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phosphorylated JUN, Mouse, Clone: C-J 4C4, Abnova™

Catalog No. 89113153 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-153 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-153 Supplier Abnova Corporation Supplier No. MAB7251
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against phosphorylated JUN.

Specifications

Antigen phosphorylated JUN
Applications Western Blot
Classification Monoclonal
Clone C-J 4C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against phosphorylated JUN.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene Alias AP-1/AP1/c-Jun
Gene Symbols JUN
Host Species Mouse
Immunogen Human/Rat JUN phosphorylated at T91 and T93.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3725/24516
Test Specificity This recognizes c-Jun phosphorylated at T91 and T93, or just at T91 but not at T93 alone. Not yet tested in other species. Predicted molecular weight: 36 KDa. Not tested in other applications.
Target Species Human, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.