Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotyrosine, Mouse, Clone: G104, Abnova™

Catalog No. 89136263 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-263 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-263 Supplier Abnova Corporation Supplier No. MAB6670
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against phosphotyrosine.

Specifications

Antigen Phosphotyrosine
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone G104
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against phosphotyrosine.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Host Species Mouse
Immunogen alanine and glycine in a 1:1:1 ratio polymerized in the presence of KLH with EDC., Phosphotyrosine
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with phosphotyrosine, and detects the presence of phosphotyrosine in proteins of both unstimulated and stimulated cell lysates. Does not cross react with phosphoserine or phosphothreonine.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.