Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotyrosine, Mouse, Clone: PY20, Abnova™

Catalog No. 89134855 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-855 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-855 Supplier Abnova Corporation Supplier No. MAB11325
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Phosphotyrosine.

Specifications

Antigen Phosphotyrosine
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PY20
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Phosphotyrosine.
Dilution Western Blot (1:200 -1:2000) Immunocytochemistry/Immunofluorescence (1:10 - 1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) Immunoprecipitation (1:10 - 1:500) The optimal working dilution should be det
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Host Species Mouse
Immunogen Phosphotyrosine was used as the immunogen.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.