Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotyrosine, Mouse, PE, Clone: P-Tyr-01, Abnova™

Catalog No. 89136780 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-780 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-780 Supplier Abnova Corporation Supplier No. MAB9815

Mouse monoclonal antibody raised against Phosphotyrosine.

Specifications

Antigen Phosphotyrosine
Applications Flow Cytometry
Classification Monoclonal
Clone P-Tyr-01
Conjugate PE
Formulation In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Phosphotyrosine conjugated with BSA.
Purification Method Size-exclusion chromatography purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.