Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Pim-1, Clone: 19F7,

Catalog No. MABC553MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC553MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC553MI Supplier MilliporeSigma™ Supplier No. MABC553
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Pim-1 clone: 19F7 in Human samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Pim-1 is a serine/threonine protein kinase encoded by the PIM1 (or PIM) gene in human. Pim-1 is expressed primarily in B-lymphoid and myeloid cell lines, and is overexpressed in hematopoietic malignancies and prostate cancer. It plays a role in signal transduction in blood cells, contributing to both cell proliferation and survival, and thus provides a selective advantage in tumorigenesis. Both the human and orthologous mouse genes have been reported to encode two isoforms (with preferential cellular localization) resulting from the use of alternative in-frame translation initiation codons, the upstream non-AUG (CUG) and downstream AUG codons (PMIDs:16186805, 1825810).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pim-1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 19F7
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P11309
Gene Symbols PIM1
Host Species Mouse
Immunogen Conjugated linear peptide corresponding to the C-terminus of human Pim-1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001230115
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.