Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIN1, Mouse, Clone: 3G8, Abnova™

Catalog No. 89109000 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-000 Supplier Abnova Corporation Supplier No. MAB1080
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant PIN1.

The human PIN1 gene encodes an essential nuclear peptidylprolyl cis-trans isomerase (PPIase; EC 5.2.1.8) involved in regulation of mitosis. PIN1 belongs to a class of PPIases that includes the E. coli parvulin, yeast Ess1, and Drosophila dodo (dod) gene products (Lu et al., 1996 [PubMed 8606777]).[supplied by OMIM

Specifications

Antigen PIN1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant PIN1.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PIN1
Gene Alias DOD/UBL5
Gene Symbols PIN1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human PIN1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5300
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.