Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5K2B, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105844 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-844 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-844 Supplier Abnova Corporation Supplier No. H00008396A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PIP5K2B.

The protein encoded by this gene catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. The encoded protein sequence does not show similarity to other kinases, but the protein does exhibit kinase activity. Additionally, the encoded protein interacts with p55 TNF receptor. [provided by RefSeq

Sequence: AYSKIKVDNHLFNKENLPSRFKFKEYCPMVFRNLRERFGIDDQDYQNSVTRSAPINSDSQGRCGTRFLTTYDRRFVIKTVSSEDVAEM

Specifications

Antigen PIP5K2B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PIP5K2B.
Formulation 50% glycerol
Gene PIP4K2B
Gene Accession No. NM_003559
Gene Alias PI5P4KB/PIP5K2B/PIP5KIIB/PIP5KIIbeta
Gene Symbols PIP4K2B
Host Species Mouse
Immunogen PIP5K2B (NP_003550, 73 a.a. ∽ 160 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8396
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.