Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITX2 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89105125 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-125 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-125 Supplier Abnova Corporation Supplier No. H00005308A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant PITX2.

This gene encodes a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. The encoded protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression. This protein plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes, is involved in the development of the eye, tooth and abdominal organs, and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. Mutations in this gene are associated with Axenfeld-Rieger syndrome, iridogoniodysgenesis syndrome, and sporadic cases of Peters anomaly. A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq

Sequence: MNCMKGPLHLEHRAAGTKLSAVSSSSCHHPQPLAMASVLAPGQPRSLDSSKHRLEVHTISDTSSPEAAEKDKSQQGKNEDVGAEDPSKKKRQRRQRTHFTSQQLQELEATFQRNRYPDMSTREEIAVWTNLTEARVRVWFKNRRAKWRKRERNQQAELCKNGFGPQFNGLMQPYDDMYPGYSYNNWAAKGLTSASLSTKSFPFFNSMNVNPLSSQSMFSPPNSISSMSMSSSMVPSAVTGVPGSSLNSLNNLNNLSSPSLNSAVPTPACPYAPPTPPYVYRDTCNSSLASLRLKAKQHSSFGYASVQNPASNLSACQYAVDRPV

Specifications

Antigen PITX2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant PITX2.
Formulation 50% glycerol
Gene PITX2
Gene Accession No. BC013998
Gene Alias ARP1/Brx1/IDG2/IGDS/IGDS2/IHG2/IRID2/MGC111022/MGC20144/Otlx2/PTX2/RGS/RIEG/RIEG1/RS
Gene Symbols PITX2
Host Species Mouse
Immunogen PITX2 (AAH13998, 1 a.a. ∽ 324 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5308
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.