Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKD1L2, Mouse, Clone: 3F6, Abnova™

Catalog No. 89107545 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-545 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-545 Supplier Abnova Corporation Supplier No. H00114780M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PKD1L2.

This gene encodes a member of the polycystin protein family. The encoded protein contains 11 transmembrane domains, a latrophilin/CL-1-like GPCR proteolytic site (GPS) domain, and a polycystin-1, lipoxygenase, alpha-toxin (PLAT) domain. This protein may function as a component of cation channel pores. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MGEDSPVAMFSWYLDNTPTEQAEPLPDACRLRGFWPRSLTLLQSNTSTLLLNSSFLQSRGEVIRIRATALTRHAYGEDTYVISTVPPREVPACTIAPEEGTVLTSFAIFCNASTALGPLEFCFCLESGSCLHCGPEPALPSVYLPLGEENNDFVLTVVISATNRAGDTQQTQAMAKVALGDTCVEDVAFQAAVSEKIPTALQGEGGPEQLLQLAKAVSSMLNQEHESQGSGQSLSIDVRQKVREHVLGSLSAVTTGLEDVQRVQELAEVLREVTCRSKELTPSAQGSCMGDSWEGAPPAAHVSHAR

Specifications

Antigen PKD1L2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant PKD1L2.
Formulation ascites with no preservative
Gene PKD1L2
Gene Accession No. BC014157
Gene Alias DKFZp686J19100/FLJ45333/PC1L2
Gene Symbols PKD1L2
Host Species Mouse
Immunogen PKD1L2 (AAH14157,1 a.a. ∽ 306 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114780
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.