Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKP3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106480 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-480 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-480 Supplier Abnova Corporation Supplier No. H00011187A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PKP3.

This gene encodes a member of the arm-repeat (armadillo) and plakophilin gene families. Plakophilin proteins contain numerous armadillo repeats, localize to cell desmosomes and nuclei, and participate in linking cadherins to intermediate filaments in the cytoskeleton. This protein may act in cellular desmosome-dependent adhesion and signaling pathways. [provided by RefSeq

Sequence: RAGGLDWPEATEVSPSRTIRAPAVRTLQRFQSSHRSRGVGGAVPGAVLEPVARAPSVRSLSLSLADSGHLPDVHGFNSYGSHRTLQRLSSGFDDIDLPSA

Specifications

Antigen PKP3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PKP3.
Formulation 50% glycerol
Gene PKP3
Gene Accession No. NM_007183
Gene Symbols PKP3
Host Species Mouse
Immunogen PKP3 (NP_009114, 225 a.a. ∽ 324 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11187
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.