Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-PL Scramblase 1, Clone: 4D2,

Catalog No. MABS483MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS483MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS483MI Supplier MilliporeSigma™ Supplier No. MABS483
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PL Scramblase 1 clone: 4D2 in Human samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting

Phospholipid scramblase 1 (UniProt O15162; also known as Ca2+-dependent phospholipid scramblase 1, Erythrocyte phospholipid scramblase, MmTRA1b, PL scramblase 1) is encoded by the PLSCR1 (also known as MMTRA1B) gene (Gene ID 5359) in human. Plasma membrane phospholipids are distributed asymmetrically between the inner and outer leaflets. Such asymmetrical distribution collapses in response to blood coagulation and apoptosis, resulting in phospholipid 'scrambling' between the two leaflets of the plasma membrane. Flippases, floppases, and scramblases are three types enzymes known to mediate transbilayer lipid motion. Flippases and floppases function via an ATP-dependent mechanism, while scramblases mediate transbilayer movement in a non-selective and energy-independent manner. Originally identified in 1996 as a 37kDa erythrocyte type II transmembrane protein that mediates calcium-dependent membrane phospholipids redistribution, PL Scramblase 1 is the protein product encoded by the founding member of the PLSCR family of genes (PLSCR1-5). All PLSCR family members, with the exception of PLSCR2, possess a proline-rich N-terminal region containing PxxP and PPxY domains, a cysteine-rich region, a conserved calcium-binding domain (EF-hand-like), and a putative transmembrane region enriched in hydrophobic amino acids. In addition, PLSCR1 contains a nuclear localization signal (NLS) and a DNA-binding domain that are essential for its nuclear localization and associated nuclear function. In addition to PLSCRs, TMEM16 and XKR family members have also been reported to mediate scramblase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PL Scramblase 1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4D2
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O15162
Gene Symbols PLSCR1
Host Species Mouse
Immunogen Recombinant protein corresponding to human PL Scramblase 1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_066928
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.