Learn More
PLA1A, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PLA1A.
Supplier: Abnova Corporation H00051365A01
Description
Phosphatidylserine-specific phospholipase A1-alpha (PLA1A) acts specifically on phosphatidylserine (PS) and 1-acyl-2-lysophosphatidylserine (lyso-PS) to hydrolyze fatty acids at the sn-1 position of these phospholipids.[supplied by OMIM
Sequence: DQPGCPTFFYAGYSYLICDHMRAVHLYISALENSCPLMAFPCASYKAFLAGRCLDCFNPFLLSCPRIGLVEQGGVKIEPLPKEVKVYLLTTSSAPYCMHHSpecifications
PLA1A | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant PLA1A. | |
PLA1A | |
PS-PLA1/PSPLA1 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_015900 | |
PLA1A | |
PLA1A (NP_056984, 241 a.a. ∽ 340 a.a) partial recombinant protein with GST tag. | |
RUO | |
51365 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only