Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLAT, Mouse, Clone: 629-2A153, Abnova™

Catalog No. 89111595 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-595 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-595 Supplier Abnova Corporation Supplier No. MAB3792
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant PLAT.

This gene encodes tissue-type plasminogen activator, a secreted serine protease which converts the proenzyme plasminogen to plasmin, a fibrinolytic enzyme. Tissue-type plasminogen activator is synthesized as a single chain which is cleaved by plasmin to a two chain disulfide linked protein. This enzyme plays a role in cell migration and tissue remodeling. Increased enzymatic activity causes hyperfibrinolysis, which manifests as excessive bleeding; decreased activity leads to hypofibrinolysis which can result in thrombosis or embolism. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Antigen PLAT
Applications ELISA, Western Blot
Classification Monoclonal
Clone 629-2A153
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PLAT.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 1mM NaCl, pH 6.6
Gene PLAT
Gene Alias DKFZp686I03148/T-PA/TPA
Gene Symbols PLAT
Host Species Mouse
Immunogen Recombinant protein corresponding to human PLAT.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5327
Test Specificity This epitope is localized to the heavy chain of human tPA and binds well to both the free and complexed form.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.