Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plat, Mouse, Clone: H27B6, Abnova™

Catalog No. 89111593 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-593 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-593 Supplier Abnova Corporation Supplier No. MAB3790
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Plat.

Specifications

Antigen Plat
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone H27B6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Plat.
Dilution Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 150mM NaCl, pH 7.4
Gene Plat
Gene Alias AU020998/AW212668/D8Ertd2e/MGC18508/t-PA/tPA
Gene Symbols Plat
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse Plat.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18791
Target Species Mouse, Rat
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.