Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plau, Mouse, Clone: B042M, Abnova™

Catalog No. 89111983 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-983 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-983 Supplier Abnova Corporation Supplier No. MAB4767

Mouse monoclonal antibody raised against Plau.

Specifications

Antigen Plau
Applications Western Blot
Classification Monoclonal
Clone B042M
Conjugate Unconjugated
Description plasminogen activator, urokinase
Formulation In 50mM sodium phosphate buffer, 150mM NaCl, pH 7.4
Gene Plau
Gene Alias u-PA, uPA
Gene Symbols Plau
Host Species Mouse
Immunogen Mouse Plau.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18792
Test Specificity Mouse Urokinase (uPA).
Target Species Mouse
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.