Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-PLGF, Clone: 3A6.2,

Catalog No. MABC527MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC527MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC527MI Supplier MilliporeSigma™ Supplier No. MABC527
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PLGF clone: 3A6.2 in Human samples, and it is validated for Western Blotting

Placenta growth factor (PlGF) (UniProt P49763) is encoded by the PGF gene in human. PlGF is a secreted protein primarily produced by placental trophoblast tissues, but is also expressed in other endothelial tissues and cancers. There are three known isoforms of PlGF (PIGF-1, PIGF-2, PIGF-3) that result due to alternative splicing. PIGF-2 is expressed up to week 8 in the placenta; while PIGF-1 and PIGF-3 are continuously expressed by the placental tissues. PIGF-1 expression is also found in colon and mammary carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLGF
Applications Western Blot
Classification Monoclonal
Clone 3A6.2
Formulation Purified mouse monoclonal IgG2bλ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P49763
Gene Symbols PGF; PGFL; PLGF
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human PLGF.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001193941
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.