Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLOD3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105984 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-984 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-984 Supplier Abnova Corporation Supplier No. H00008985A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant PLOD3.

The protein encoded by this gene is a membrane-bound homodimeric enzyme that is localized to the cisternae of the rough endoplasmic reticulum. The enzyme (cofactors iron and ascorbate) catalyzes the hydroxylation of lysyl residues in collagen-like peptides. The resultant hydroxylysyl groups are attachment sites for carbohydrates in collagen and thus are critical for the stability of intermolecular crosslinks. Some patients with Ehlers-Danlos syndrome type VIB have deficiencies in lysyl hydroxylase activity. [provided by RefSeq

Sequence: MTSSGPGPRFLLLLPLLLPPAASASDRPRGRDPVNPEKLLVITVATAETEGYLRFLRSAEFFNYTVRTLGLGEEWRGGDVARTVGGGQKVRWLKKEMEKYADREDMIIMFVDSYDVILAGSPTELLKKFVQSGSRLLFSAESFCWPEWGLAEQYPEVGTGKRFLNSGGFIGFATTIHQIVRQWKYKDDDDDQLFYTRLYLDPGLREKLSLNLDHKSRIFQNLNGALDEVVLKFDRNRVRIRNVAYDTLPIVVHGNGPTKLQLNYLGNYVPNGWTPEGGCGFCNQDRRTLPGGQPPPRVFLAVFVEQPTPFLPRFLQRLLLLDYPPDRVTLFLHNNEVFHEPHIADSWPQLQDHFSAVKLVGPEEALSPGEARDMAMDLCRQDPECEFYFSLDADAVLTNLQTLRILIEENRKVIAPMLSRHGKLWSNFWGALSPDEYYARSEDYVELVQRKRVGVWNVPYISQAYVIRGDTLRMELPQRDVFSGSDTDPDMAFCKSFRDKGIFLHLSNQHEFGRLLATSRYDTEHLHPDLWQIFDNPVDWKEQYIHENYSRALEGEGIVEQPCPDVYWFPLLSEQMCDELVAEMEHYGQWSGGRHEDSRLAGGYENVPTVDIHMKQVGYEDQWLQLLRTYVGPMTESLFPGYHTKARAVMNFVVRYRPDEQPSLRPHHDSSTFTLNVALNHKGLDYEGGGCRFLRYDCVISSPRKGWALLHPGRLTHYHEGLPTTWGTRYIMVSFVDP

Specifications

Antigen PLOD3
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant PLOD3.
Formulation 50% glycerol
Gene PLOD3
Gene Accession No. BC011674
Gene Alias LH3
Gene Symbols PLOD3
Host Species Mouse
Immunogen PLOD3 (AAH11674.1, 1 a.a. ∽ 738 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8985
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.