Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plscr1, Mouse, Clone: 13A6 [TEC-23], Abnova™

Catalog No. 89112893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-893 Supplier Abnova Corporation Supplier No. MAB6482
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Plscr1.

Specifications

Antigen Plscr1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 13A6 [TEC-23]
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Plscr1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In ascites (0.09% sodium azide)
Gene Plscr1
Gene Alias MGC93252
Gene Symbols Plscr1
Host Species Mouse
Immunogen BALB/c mice were immunized with pooled lipid rafts isolated from RBL-2H3 cells. Hybridoma cells were obtained after fusion of SP02 mouse myeloma cells with spleen cells of immunized mice using standard procedures.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117540
Test Specificity This antibody binds to rat Phospholipid Scramblase 1 (PLSCR1), an 37-49 KDa membrane-associated protein accelerating bidirectional movement of plasmamembrane phospholipids during conditions of elevated calcium.
Target Species Rat
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.