Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLXDC1, Mouse, Clone: 197C193 (IM193), Abnova™

Catalog No. 89107868 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-868 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-868 Supplier Abnova Corporation Supplier No. MAB0098
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of PLXDC1.

Specifications

Antigen PLXDC1
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 197C193 (IM193)
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of PLXDC1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene PLXDC1
Gene Alias DKFZp686F0937/FLJ36270/FLJ45632/TEM3/TEM7
Gene Symbols PLXDC1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 411-427 of human PLXDC1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57125
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.