Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMA, Mouse, Clone: YPMA-2, Abnova™

Catalog No. 89136193 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-193 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-193 Supplier Abnova Corporation Supplier No. MAB6363
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native PMA.

Specifications

Antigen PMA
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone YPMA-2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native PMA.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Host Species Mouse
Immunogen Native purified human prostate mucin tissues PMA.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts specifically with prostate mucin antigen.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.