Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMA1, Mouse, Clone: 40B7, Abnova™

Catalog No. 89111366 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-366 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-366 Supplier Abnova Corporation Supplier No. MAB3349
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native PMA1.

Specifications

Antigen PMA1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 40B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native PMA1.
Dilution Western Blot (1:500-1:5000) Immunocytochemistry (1:250-1:2500) Immunofluorescence (1:250-1:2500) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.09% sodium azide)
Gene PMA1
Gene Symbols PMA1
Host Species Mouse
Immunogen Native purified PMA1 from yeast plasma membrane preparations.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 852876
Target Species Yeast
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.