Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMPCB, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106094 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-094 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-094 Supplier Abnova Corporation Supplier No. H00009512A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PMPCB.

This gene is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex. [provided by RefSeq

Sequence: IQNSTLGEAEIERERGVILREMQEVETNLQEVVFDYLHATAYQNTALGRTILGPTENIKSISRKDLVDYITTHYKGPRIVLAAAGGVSHDELLDLAKFHFGDSLCTHKG

Specifications

Antigen PMPCB
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PMPCB.
Formulation 50% glycerol
Gene PMPCB
Gene Accession No. NM_004279
Gene Alias Beta-MPP/MPP11/MPPB/MPPP52
Gene Symbols PMPCB
Host Species Mouse
Immunogen PMPCB (NP_004270, 161 a.a. ∽ 269 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9512
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.