Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105155 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-155 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-155 Supplier Abnova Corporation Supplier No. H00005378A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PMS1.

This gene encodes a protein belonging to the DNA mismatch repair mutL/hexB family. This protein is thought to be involved in the repair of DNA mismatches, and it can form heterodimers with MLH1, a known DNA mismatch repair protein. Mutations in this gene cause hereditary nonpolyposis colorectal cancer type 3 (HNPCC3) either alone or in combination with mutations in other genes involved in the HNPCC phenotype, which is also known as Lynch syndrome. [provided by RefSeq

Sequence: KELIENSLDAGATSVDVKLENYGFDKIEVRDNGEGIKAVDAPVMAMKYYTSKINSHEDLENLTTYGFRGEALGSICCIAEVLITTRTAADNFSTQYVLDGSGHILSQKPS

Specifications

Antigen PMS1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PMS1.
Formulation 50% glycerol
Gene PMS1
Gene Accession No. NM_000534
Gene Alias DKFZp781M0253/FLJ98259/HNPCC3/PMSL1/hPMS1
Gene Symbols PMS1
Host Species Mouse
Immunogen PMS1 (NP_000525, 26 a.a. ∽ 135 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5378
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.