Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS2, Mouse, Clone: 349.29.5.2, Abnova™

Catalog No. 89110816 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-816 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-816 Supplier Abnova Corporation Supplier No. MAB2089
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant PMS2.

This gene is one of the PMS2 gene family members found in clusters on chromosome 7. The product of this gene is involved in DNA mismatch repair. It forms a heterodimer with MLH1 and this complex interacts with other complexes bound to mismatched bases. Mutations in this gene are associated with hereditary nonpolyposis colorectal cancer, Turcot syndrome, and are a cause of supratentorial primitive neuroectodermal tumors. Alternatively spliced transcript variants have been observed for this gene. [provided by RefSeq

Specifications

Antigen PMS2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1204
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant PMS2.
Dilution ELISA (1:5000-1:20000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene PMS2
Gene Alias HNPCC4/PMS2CL/PMSL2
Gene Symbols PMS2
Host Species Mouse
Immunogen Recombinant protein corresponding to the first 133 amino acids of human PMS2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5395
Test Specificity This is an protein A purified antibody directed against PMS2-134 and reacts with full length version of PMS2 in human and hamster tissues. The epitope was putatively mapped to amino acids 58-81 of human PMS2.
Target Species Chimpanzee, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.