Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNLIP Mouse anti-Human, Monoclonal Antibody, Abnova™

Catalog No. 89110892 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-892 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-892 Supplier Abnova Corporation Supplier No. MAB2206
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native PNLIP.

This gene is a member of the lipase gene family. It encodes a carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides, and is essential for the efficient digestion of dietary fats. This gene is expressed specifically in the pancreas. [provided by RefSeq

Specifications

Antigen PNLIP
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native PNLIP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene PNLIP
Gene Alias PL
Gene Symbols PNLIP
Host Species Mouse
Immunogen Native purified PNLIP from human pancreas.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5406
Test Specificity These antibodies specifically recognize pancreatic lipase.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.