Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPO, Mouse, Clone: AT2C7, Abnova™

Catalog No. 89109303 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-303 Supplier Abnova Corporation Supplier No. MAB11110
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant PNPO.

The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5'-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine. Mutations in this gene result in pyridoxamine 5'-phosphate oxidase (PNPO) deficiency, a form of neonatal epileptic encephalopathy. [provided by RefSeq]

Specifications

Antigen PNPO
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT2C7
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant PNPO.
Dilution Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PNPO
Gene Alias FLJ10535/PDXPO
Gene Symbols PNPO
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 57-261 of human PNPO.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55163
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.