Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLD3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106388 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-388 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-388 Supplier Abnova Corporation Supplier No. H00010714A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant POLD3.

The DNA polymerase delta complex is involved in DNA replication and repair, and it consists of the proliferating cell nuclear antigen (PCNA; MIM 176740), the multisubunit replication factor C (see MIM 102579), and the 4 subunit polymerase complex: POLD1 (MIM 174761), POLD2 (MIM 600815), POLD3, and POLD4 (MIM 611525) (Liu and Warbrick, 2006 [PubMed 16934752]).[supplied by OMIM

Sequence: PPLEPVPKTEPEPPSVKSSSGENKRKRKRVLKSKTYLDGEGCIVTEKVYESESCTDSEEELNMKTSSVHRPPAMTVKKEPREERKGPKKGTAALGKANRQVSITGFFQRK

Specifications

Antigen POLD3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant POLD3.
Formulation 50% glycerol
Gene POLD3
Gene Accession No. NM_006591
Gene Alias KIAA0039/MGC119642/MGC119643/P66/P68
Gene Symbols POLD3
Host Species Mouse
Immunogen POLD3 (NP_006582, 357 a.a. ∽ 466 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10714
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.