Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLE, Mouse, Clone: 93H3A, Abnova™

Catalog No. 89111420 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-420 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-420 Supplier Abnova Corporation Supplier No. MAB3438

Mouse monoclonal antibody raised against partial recombinant POLE.

Specifications

Antigen POLE
Applications Immunoprecipitation
Classification Monoclonal
Clone 93H3A
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant POLE.
Dilution Immunoprecipitation (2 ug/mg of protein lysate) Western Blot (2-4 ug/mL The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene POLE
Gene Alias DKFZp434F222/FLJ21434/POLE1
Gene Symbols POLE
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-176 of human POLE.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5426
Test Specificity This antibody recognizes both polε and polε*. Does not cross-react with DNA polymerase alpha or δ and does not neutralize the polymerase or exonuclease activity of the enzyme.
Target Species Bovine, Human, Monkey, Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.